Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002087.3 | NP_002078.1 | 13 | 2279 | ||
XM_005257253.1 | XP_005257310.1 | 13 | 2324 | |||
GenBank mRNA | AF055008.1 | - | 13 | 2085 | ||
AK000607.1 | - | 13 | 2063 | |||
AK023348.1 | - | 10 | 1567 | |||
AK222522.1 | - | 13 | 2063 | |||
AK303830.1 | - | 10 | 1589 | |||
AY124489.1 | - | 12 | 2047 | |||
BC000324.2 | - | 12 | 1777 | 2821810 | ||
BC010577.2 | - | 12 | 2033 | 3457813 | ||
M75161.1 | - | 12 | 2035 | |||
X62320.1 | - | 13 | 2063 |