Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172129.1 | NP_001165600.1 | 13 | 1753 | ||
NM_001172130.1 | NP_001165601.1 | 13 | 1750 | |||
NM_001172131.1 | NP_001165602.1 | 13 | 1750 | |||
NM_001172132.1 | NP_001165603.1 | 14 | 1819 | |||
NM_001172133.1 | NP_001165604.1 | 14 | 1817 | |||
NM_002110.3 | NP_002101.2 | 13 | 1753 | |||
GenBank mRNA | AK026432.1 | - | 13 | 1687 | ||
AK225819.1 | - | 13 | 1699 | |||
AK289896.1 | - | 13 | 1741 | |||
AK290928.1 | - | 14 | 1765 | |||
AK298726.1 | - | 14 | 1763 | |||
AK314335.1 | - | 13 | 1666 | |||
BC014435.1 | - | 13 | 1639 | 4855747 | ||
BC094847.1 | - | 13 | 1650 | 5923744 | ||
BC108930.1 | - | 13 | 1680 | 40025497 | ||
BC108931.1 | - | 13 | 1680 | 40025500 | ||
BC113854.1 | - | 13 | 1677 | 40025503 | ||
BC114463.1 | - | 13 | 1680 | 40025502 | ||
M16592.1 | - | 12 | 1528 |