Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002158.3 | NP_002149.2 | 7 | 1433 | ||
XM_005264282.2 | XP_005264339.1 | 6 | 1307 | |||
XM_005264283.2 | XP_005264340.1 | 8 | 1628 | |||
XM_006712001.3 | XP_006712064.1 | 7 | 1364 | |||
XM_006712002.3 | XP_006712065.1 | 6 | 1324 | |||
XM_011532808.2 | XP_011531110.1 | 8 | 1550 | |||
XM_017003956.1 | XP_016859445.1 | 7 | 1768 | |||
XM_017003957.1 | XP_016859446.1 | 8 | 1625 | |||
XM_017003958.1 | XP_016859447.1 | 8 | 1624 | |||
XM_017003959.1 | XP_016859448.1 | 8 | 1471 | |||
GenBank mRNA | BC063305.1 | - | 7 | 1358 | 30343577 | |
BC069731.1 | - | 5 | 926 | |||
U57029.1 | - | 5 | 1129 |