Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002175.2 | NP_002166.2 | 1 | 668 | ||
GenBank mRNA | BC069329.1 | - | 1 | 628 | 7262150 | |
BC069372.1 | - | 1 | 628 | 7262162 | ||
BC069408.1 | - | 1 | 628 | 7262174 | ||
BC096699.1 | - | 1 | 662 | 40015057 | ||
BC101638.1 | - | 1 | 627 | 8069144 | ||
BC101640.1 | - | 1 | 627 | 8069146 | ||
M12350.1 | - | 1 | 668 | |||
M28586.1 | - | 1 | 668 | |||
V00540.1 | - | 1 | 627 | |||
X00145.1 | - | 1 | 668 |
Gene Symbol: IFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | E00175.1 | - | 1 | 604 |