Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002213.4 | NP_002204.2 | 4 | 831 | ||
XM_005247436.2 | XP_005247493.1 | 4 | 1064 | |||
XM_006713630.2 | XP_006713693.1 | 4 | 1064 | |||
XM_017006351.1 | XP_016861840.1 | 4 | 602 | |||
XM_017006352.1 | XP_016861841.1 | 4 | 541 | |||
XM_017006353.1 | XP_016861842.1 | 4 | 573 | |||
XM_017006354.1 | XP_016861843.1 | 4 | 1064 | |||
XM_017006355.1 | XP_016861844.1 | 4 | 541 | |||
GenBank mRNA | AK315429.1 | - | 4 | 567 | ||
BC006541.2 | - | 4 | 823 | 2958666 | ||
J05633.1 | - | 4 | 572 | |||
M35011.1 | - | 4 | 555 | |||
X53002.1 | - | 4 | 862 |