Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004507.3 | NP_004498.1 | 3 | 309 | ||
NR_037917.1 | 3 | 367 | ||||
XM_017012115.1 | XP_016867604.1 | 3 | 307 | |||
GenBank mRNA | AF076844.1 | - | 3 | 266 | ||
AF110393.1 | - | 3 | 263 | |||
AK290459.1 | - | 3 | 246 | |||
AK294117.1 | - | 3 | 295 | |||
BC007013.1 | - | 3 | 227 | 3684398 | ||
BG286955.1 | - | 337 | ||||
BT019481.1 | - | 3 | 203 | |||
BT019482.1 | - | 3 | 203 | |||
CR536552.1 | - | 3 | 203 | |||
Y16893.1 | - | 3 | 203 |