Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286099.1 | NP_001273028.1 | 3 | 1589 | ||
NM_005284.4 | NP_005275.1 | 2 | 1480 | |||
GenBank mRNA | AK295250.1 | - | 3 | 1534 | ||
BC043414.2 | - | 1 | 1268 | 5294842 |
Set Membership: |
![]() |