Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGFG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006076.4 | NP_006067.3 | 12 | 2134 | ||
XM_005250306.2 | XP_005250363.1 | 13 | 2153 | |||
XM_017012112.1 | XP_016867601.1 | 12 | 2075 | |||
XM_017012114.1 | XP_016867603.1 | 11 | 1833 | |||
GenBank mRNA | BC017329.2 | - | 12 | 2086 | 4810320 |