Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFTUD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142605.1 | NP_001136077.1 | 9 | 1012 | ||
NM_001258353.1 | NP_001245282.1 | 10 | 1258 | |||
NM_001258354.1 | NP_001245283.1 | 10 | 1091 | |||
NM_004247.3 | NP_004238.3 | 10 | 1121 | |||
GenBank mRNA | AJ505017.1 | - | 6 | 495 | ||
AK126464.1 | - | 11 | 927 | |||
AK128148.1 | - | 10 | 1258 | |||
AK225335.1 | - | 10 | 881 | |||
AK293108.1 | - | 10 | 884 | |||
AK296367.1 | - | 9 | 815 | |||
AK297392.1 | - | 10 | 897 | |||
AK302828.1 | - | 10 | 761 | |||
AK316098.1 | - | 9 | 832 | |||
BC002360.2 | - | 10 | 891 | 2960986 | ||
CR456774.1 | - | 9 | 860 | |||
D21163.1 | - | 10 | 920 |