Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021262.1 | XP_016876751.1 | 2 | 408 | ||
XM_017021263.1 | XP_016876752.1 | 3 | 252 | |||
XM_017021264.1 | XP_016876753.1 | 2 | 324 | |||
GenBank mRNA | AF279372.1 | - | 3 | 272 | ||
AK293515.1 | - | 3 | 327 | |||
BC003622.2 | - | 4 | 432 | 3010239 | ||
BC007428.2 | - | 4 | 432 | 3010239 | ||
BC018192.1 | - | 4 | 381 | 3867269 | ||
BC037305.2 | - | 2 | 298 | |||
BQ434653.1 | - | 4 | 460 | |||
BX420122.2 | - | 4 | 496 | |||
CB997740.1 | - | 350 | ||||
U51336.1 | - | 4 | 325 |