Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: INCENP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040694.1 | NP_001035784.1 | 19 | 3517 | ||
NM_020238.2 | NP_064623.2 | 18 | 3505 | |||
XM_006718533.2 | XP_006718596.1 | 19 | 3491 | |||
XM_011544995.2 | XP_011543297.1 | 19 | 3524 | |||
XM_011544996.2 | XP_011543298.1 | 19 | 3512 | |||
XM_011544997.2 | XP_011543299.1 | 18 | 3472 | |||
XM_011544998.2 | XP_011543300.1 | 18 | 3500 | |||
GenBank mRNA | AF282265.1 | - | 17 | 3318 |