Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ICA1 SIRNA Assays ›
Gene Symbol:
ICA1
Gene Name:
islet cell autoantigen 1
Gene Aliases:
ICA69, ICAp69
Chromosome Location:
Chr.7: 8113184 - 8262687 on Build GRCh38
UniGene ID:
Hs.487561
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
145392
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ICA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136020.2
NP_001129492.1
14
1696
NM_001276478.1
NP_001263407.1
14
1678
NM_004968.3
NP_004959.2
14
1681
NM_022307.2
NP_071682.1
14
1496
XM_005249735.3
XP_005249792.1
14
1618
XM_011515345.1
XP_011513647.1
15
1783
XM_011515346.1
XP_011513648.1
15
1768
XM_011515347.2
XP_011513649.1
15
1655
XM_011515348.1
XP_011513650.1
15
1701
XM_011515349.1
XP_011513651.1
15
1634
XM_011515350.1
XP_011513652.1
15
1705
XM_011515351.1
XP_011513653.1
15
1840
XM_011515352.1
XP_011513654.1
15
1659
XM_011515353.2
XP_011513655.1
15
1712
XM_011515354.1
XP_011513656.1
12
1296
XM_011515355.2
XP_011513657.1
10
1189
XM_011515356.2
XP_011513658.1
10
1143
XM_011515357.2
XP_011513659.1
10
997
XM_017012116.1
XP_016867605.1
15
1551
XM_017012117.1
XP_016867606.1
14
1654
XM_017012118.1
XP_016867607.1
14
1614
XM_017012119.1
XP_016867608.1
14
1589
XM_017012120.1
XP_016867609.1
14
1635
XM_017012121.1
XP_016867610.1
14
1502
XM_017012122.1
XP_016867611.1
15
1881
XM_017012123.1
XP_016867612.1
15
1820
XM_017012124.1
XP_016867613.1
15
1981
XM_017012125.1
XP_016867614.1
15
1731
GenBank mRNA
AK292106.1
-
14
1565
BC008640.2
-
14
1613
3854722
EU416199.1
-
14
1516
L01100.1
-
14
1607
L21181.1
-
14
1505
U26591.1
-
14
1496
U26592.1
-
15
1767
U37183.1
-
12
1395
U38260.1
-
9
1243
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_030844, XM_006236092, XM_006236095, XM_006236094, XM_008762728, and others.
Gene Ontology Categories:
Function(s)
Process(es)
neurotransmitter transport
regulation of protein homodimerization activity
regulation of insulin secretion
molecular_function
protein binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f4tdd:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0