Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004501.3 | NP_004492.2 | 11 | 2101 | ||
NM_031844.2 | NP_114032.2 | 11 | 2158 | |||
XM_017001115.1 | XP_016856604.1 | 11 | 2353 | |||
XM_017001116.1 | XP_016856605.1 | 11 | 2158 | |||
XM_017001117.1 | XP_016856606.1 | 11 | 2147 | |||
GenBank mRNA | AF068846.1 | - | 11 | 2154 | ||
AF461014.1 | - | 8 | 1679 | |||
AK126868.1 | - | 12 | 1899 | |||
AK130669.1 | - | 10 | 1207 | |||
AK297119.1 | - | 12 | 1992 | |||
BC003367.1 | - | 11 | 2032 | 2900876 | ||
BC003621.2 | - | 11 | 2034 | 2966453 | ||
BC007950.2 | - | 11 | 1582 | |||
BC015782.1 | - | 11 | 1635 | 4857112 | ||
BC024767.1 | - | 11 | 2034 | 4993703 | ||
BC034925.1 | - | 8 | 1025 | |||
X65488.1 | - | 11 | 1924 |