Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIVM-ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204425.1 | NP_001191354.1 | 22 | 4404 | ||
GenBank mRNA | AB209312.1 | - | 22 | 4196 | ||
LM994822.1 | - | 22 | 4282 |
Gene Symbol: ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000123.3 | NP_000114.2 | 14 | 3346 | ||
GenBank mRNA | AF462447.1 | - | 14 | 3303 | ||
AK294708.1 | - | 14 | 3275 | |||
BC031522.2 | - | 14 | 3332 | 3869893 | ||
BX647399.1 | - | 14 | 3175 | |||
D16305.1 | - | 14 | 2927 | |||
L20046.1 | - | 14 | 2968 | |||
LM994821.1 | - | 10 | 2416 | |||
X69978.1 | - | 14 | 3117 |