Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001226.3 | NP_001217.2 | 7 | 1633 | ||
NM_032992.2 | NP_116787.1 | 4 | 1366 | |||
NR_133012.1 | 6 | 1556 | ||||
GenBank mRNA | U20537.1 | - | 4 | 1285 |