Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287593.1 | NP_001274522.1 | 6 | 1118 | ||
NM_001287594.1 | NP_001274523.1 | 6 | 658 | |||
NM_001288.4 | NP_001279.2 | 5 | 809 | |||
GenBank mRNA | AF034607.1 | - | 5 | 814 | ||
AF109197.1 | - | 5 | 599 | |||
AK129773.1 | - | 5 | 554 | |||
AK223379.1 | - | 5 | 782 | |||
AK314014.1 | - | 5 | 554 | |||
BC064527.1 | - | 6 | 1118 | 5585323 | ||
BC095469.1 | - | 5 | 744 | 30557606 | ||
CB124154.1 | - | 3 | 346 | |||
CR542071.1 | - | 5 | 545 | |||
CX783058.1 | - | 6 | 656 | |||
U93205.1 | - | 5 | 763 | |||
X87689.1 | - | 5 | 749 |