Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290223.1 | NP_001277152.1 | 3 | 266 | ||
NM_001380.4 | NP_001371.1 | 3 | 266 | |||
XM_011539422.2 | XP_011537724.1 | 3 | 244 | |||
XM_011539424.1 | XP_011537726.1 | 3 | 154 | |||
XM_017015813.1 | XP_016871302.1 | 3 | 239 | |||
XM_017015814.1 | XP_016871303.1 | 3 | 238 | |||
XM_017015815.1 | XP_016871304.1 | 3 | 261 | |||
XM_017015817.1 | XP_016871306.1 | 3 | 238 | |||
GenBank mRNA | BC146857.1 | - | 3 | 219 | 9056785 | |
BX482033.1 | - | 3 | 221 | |||
D50857.1 | - | 3 | 180 | |||
DA350224.1 | - | 3 | 267 |