Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S1PR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320730.1 | NP_001307659.1 | 2 | 516 | ||
NM_001400.4 | NP_001391.2 | 2 | 624 | |||
GenBank mRNA | AF022137.1 | - | 1 | 110 | ||
AF233365.1 | - | 1 | 110 | |||
AK297005.1 | - | 2 | 362 | |||
AK312493.1 | - | 2 | 354 | |||
BC018650.1 | - | 2 | 358 | 4071217 | ||
BI598518.1 | - | 2 | 363 | |||
BP236900.1 | - | 2 | 352 | |||
CR541786.1 | - | 1 | 110 | |||
CR542269.1 | - | 1 | 110 | |||
CR977007.1 | - | 2 | 484 | |||
M31210.1 | - | 2 | 360 |
Set Membership: |
![]() |