Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001415.3 | NP_001406.1 | 12 | 1574 | ||
GenBank mRNA | AK290397.1 | - | 12 | 1566 | ||
BC019906.1 | - | 12 | 1574 | 4419438 |