Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ESRRG SIRNA Assays ›
Gene Symbol:
ESRRG
Gene Name:
estrogen related receptor gamma
Gene Aliases:
ERR3, ERRgamma, NR3B3
Chromosome Location:
Chr.1: 216503246 - 217137757 on Build GRCh38
UniGene ID:
Hs.444225
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
145913
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ESRRG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134285.2
NP_001127757.1
6, 7
945
NM_001243505.1
NP_001230434.1
4, 5
385
NM_001243506.1
NP_001230435.1
4, 5
455
NM_001243507.1
NP_001230436.1
5, 6
1232
NM_001243509.1
NP_001230438.1
6, 7
1323
NM_001243510.2
NP_001230439.1
6, 7
939
NM_001243511.2
NP_001230440.1
5, 6
821
NM_001243512.1
NP_001230441.1
6, 7
888
NM_001243513.1
NP_001230442.1
5, 6
908
NM_001243514.1
NP_001230443.1
4, 5
1065
NM_001243515.1
NP_001230444.1
6, 7
1164
NM_001243519.1
NP_001230448.1
6, 7
1168
NM_001438.3
NP_001429.2
4, 5
869
NM_206594.2
NP_996317.1
6, 7
989
NM_206595.2
NP_996318.1
5, 6
871
XM_011509266.2
XP_011507568.1
6, 7
1326
XM_017000626.1
XP_016856115.1
6, 7
1063
XM_017000627.1
XP_016856116.1
6, 7
1126
XM_017000638.1
XP_016856127.1
7, 8
1127
XM_017000639.1
XP_016856128.1
6, 7
967
XM_017000640.1
XP_016856129.1
5, 6
3230
XM_017000641.1
XP_016856130.1
5, 6
1025
XM_017000642.1
XP_016856131.1
6, 7
1123
XM_017000643.1
XP_016856132.1
7, 8
1120
XM_017000644.1
XP_016856133.1
6, 7
998
XM_017000645.1
XP_016856134.1
5, 6
880
XM_017000646.1
XP_016856135.1
6, 7
3363
XM_017000647.1
XP_016856136.1
5, 6
1112
XM_017000648.1
XP_016856137.1
5, 6
1001
XM_017000649.1
XP_016856138.1
7, 8
1449
GenBank mRNA
AB020639.1
-
4, 5
850
AB362218.1
-
6, 7
1242
AF058291.1
-
5, 6
845
AF094518.1
-
4, 5
821
AK131193.1
-
5, 6
1193
AK290945.1
-
6, 7
1145
AK291028.1
-
5, 6
846
AK291647.1
-
4, 5
842
AK298851.1
-
4, 5
422
AY388456.1
-
4, 5
824
AY388457.1
-
5, 6
857
AY388458.1
-
6, 7
883
AY388459.1
-
5, 6
900
AY388460.1
-
4, 5
740
AY388461.1
-
4, 5
1114
BC008218.2
-
6, 7
1028
BM723962.1
-
4, 5
385
BX647076.1
-
6, 7
945
CR749497.1
-
6, 7
938
HQ692853.1
-
4, 5
696
HQ692854.1
-
3, 4
627
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_203336, XM_006250443, AB126963, AB126962, AY341057.This siRNA also targets the following transcripts in Mouse: NM_001243792, NM_011935, BC082324, AB221580, AK140676, and others.
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
steroid hormone mediated signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
steroid hormone receptor activity
retinoic acid receptor activity
steroid binding
protein binding
zinc ion binding
AF-2 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0