Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014437.2 | NP_001014437.1 | 9 | 1057 | ||
NM_001194997.1 | NP_001181926.1 | 9 | 1057 | |||
NM_001751.5 | NP_001742.1 | 8 | 808 | |||
NM_139273.3 | NP_644802.1 | 8 | 808 | |||
NR_036542.1 | 9 | 1104 | ||||
XM_006718341.3 | XP_006718404.1 | 8 | 831 | |||
XM_017018389.1 | XP_016873878.1 | 8 | 831 | |||
XM_017018390.1 | XP_016873879.1 | 9 | 1071 | |||
XM_017018391.1 | XP_016873880.1 | 6 | 1055 | |||
XM_017018392.1 | XP_016873881.1 | 8 | 993 | |||
GenBank mRNA | AB208814.1 | - | 8 | 784 | ||
AF288206.1 | - | 8 | 741 | |||
AF288207.1 | - | 8 | 741 | |||
AK096313.1 | - | 9 | 1071 | |||
AK222614.1 | - | 8 | 749 | |||
AK289432.1 | - | 8 | 781 | |||
AK295371.1 | - | 8 | 699 | |||
AK296763.1 | - | 9 | 922 | |||
AK298514.1 | - | 9 | 1024 | |||
AK302644.1 | - | 9 | 1024 | |||
AK308005.1 | - | 9 | 977 | |||
BC002880.2 | - | 8 | 794 | 3940167 | ||
BT009913.1 | - | 8 | 724 | |||
BX647906.1 | - | 9 | 1031 | |||
L06845.1 | - | 6 | 595 |