Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001795.4 | NP_001786.2 | 12 | 3246 | ||
XM_011522801.1 | XP_011521103.1 | 12 | 3250 | |||
GenBank mRNA | AB209908.1 | - | 12 | 3146 | ||
X79981.1 | - | 11 | 3098 |