- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: CHML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001821.3 | NP_001812.2 | 1 | 2061 | ||
GenBank mRNA | AK314125.1 | - | 1 | 2325 | ||
BC117360.1 | - | 1 | 1931 | 40125911 | ||
X64728.1 | - | 1 | 2061 |