Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001824.4 | NP_001815.2 | 8 | 1633 | ||
XM_017026240.1 | XP_016881729.1 | 7 | 1518 | |||
GenBank mRNA | AJ572902.1 | - | 1 | 395 | ||
AK129878.1 | - | 8 | 1531 | |||
AK300730.1 | - | 8 | 1421 | |||
BC007462.1 | - | 8 | 1531 | 3934232 |