Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001841.2 | NP_001832.1 | 2 | 336 | ||
XM_011540629.2 | XP_011538931.1 | 2 | 619 | |||
XM_017000261.1 | XP_016855750.1 | 5 | 1311 | |||
GenBank mRNA | AJ430063.1 | - | 1 | 245 | ||
AM156854.1 | - | 1 | 209 | |||
AM156855.1 | - | 1 | 209 | |||
AM156856.1 | - | 1 | 209 | |||
BC069722.1 | - | 2 | 278 | 7262528 | ||
BC074767.2 | - | 1 | 237 | 30915186 | ||
BC095545.1 | - | 1 | 206 | |||
EU517121.1 | - | 3 | 468 | |||
X74328.1 | - | 2 | 335 |
Set Membership: |
![]() |