Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELAVL2 SIRNA Assays ›
Gene Symbol:
ELAVL2
Gene Name:
ELAV like neuron-specific RNA binding protein 2
Gene Aliases:
HEL-N1, HELN1, HUB
Chromosome Location:
Chr.9: 23690099 - 23850757 on Build GRCh38
UniGene ID:
Hs.166109
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14629
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELAVL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001171195.1
NP_001164666.1
2
420
NM_001171197.1
NP_001164668.1
2
433
NM_004432.3
NP_004423.2
2
439
XM_005251393.3
XP_005251450.1
2
577
XM_005251394.3
XP_005251451.1
2
734
XM_005251395.3
XP_005251452.1
2
514
XM_006716734.3
XP_006716797.1
3
826
XM_006716735.3
XP_006716798.1
3
294
XM_006716736.3
XP_006716799.1
2
715
XM_011517774.2
XP_011516076.1
3
840
XM_011517776.2
XP_011516078.1
2
270
XM_011517777.2
XP_011516079.1
3
826
XM_011517778.2
XP_011516080.2
2
363
XM_011517779.2
XP_011516081.1
3
338
XM_011517780.2
XP_011516082.1
3
667
XM_011517783.2
XP_011516085.1
3
700
XM_011517784.2
XP_011516086.1
2
395
XM_011517785.2
XP_011516087.1
2
200
XM_011517786.2
XP_011516088.1
2
574
XM_017014408.1
XP_016869897.1
4
841
XM_017014409.1
XP_016869898.1
3
479
XM_017014410.1
XP_016869899.1
3
623
XM_017014411.1
XP_016869900.1
3
806
XM_017014412.1
XP_016869901.1
4
788
XM_017014413.1
XP_016869902.1
4
787
XM_017014414.1
XP_016869903.1
3
810
XM_017014415.1
XP_016869904.1
2
256
XM_017014416.1
XP_016869905.1
3
329
XM_017014417.1
XP_016869906.1
3
700
XM_017014418.1
XP_016869907.1
2
534
XM_017014419.1
XP_016869908.1
2
425
XM_017014420.1
XP_016869909.1
2
428
XM_017014421.1
XP_016869910.1
2
546
XM_017014422.1
XP_016869911.1
2
715
XM_017014423.1
XP_016869912.1
2
372
XM_017014424.1
XP_016869913.1
2
718
XM_017014425.1
XP_016869914.1
2
526
GenBank mRNA
AB209294.1
-
2
439
BC030692.1
-
2
437
4826082
BC042393.1
-
2
423
Back To Top
More Information
Panther Classification:
Molecular Function -
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
regulation of transcription, DNA-templated
nucleotide binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-tbml7:80/10.239.166.229:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0