Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004386.2 | NP_004377.2 | 15 | 5601 | ||
GenBank mRNA | AB210004.1 | - | 15 | 5545 | ||
AF026547.1 | - | 14 | 5502 | |||
AK126639.1 | - | 4 | 3697 |