Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FARSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004461.2 | NP_004452.1 | 1, 2 | 203 | ||
GenBank mRNA | AF042347.1 | - | 1, 2 | 155 | ||
AK092093.1 | - | 1, 2 | 170 | |||
AK130195.1 | - | 1, 2 | 174 | |||
AK304587.1 | - | 1, 2 | 175 | |||
BC006495.2 | - | 1, 2 | 158 | 2823551 | ||
BC043565.1 | - | 1, 2 | 153 | |||
BT007198.1 | - | 1, 2 | 155 | |||
CR456740.1 | - | 1, 2 | 155 | |||
D84471.1 | - | 1, 2 | 158 | |||
U07424.1 | - | 1, 2 | 167 |