Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DNM1 SIRNA Assays ›
Gene Symbol:
DNM1
Gene Name:
dynamin 1
Gene Aliases:
DNM, EIEE31
Chromosome Location:
Chr.9: 128203355 - 128255248 on Build GRCh38
UniGene ID:
Hs.522413
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146450
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DNM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005336.2
NP_001005336.1
11, 12
1546
NM_001288737.1
NP_001275666.1
11, 12
1546
NM_001288738.1
NP_001275667.1
11, 12
1546
NM_001288739.1
NP_001275668.1
11, 12
1546
NM_004408.3
NP_004399.2
11, 12
1546
XM_005251763.2
XP_005251820.1
11, 12
1546
XM_005251768.2
XP_005251825.1
11, 12
1546
XM_005251769.2
XP_005251826.1
11, 12
1546
XM_006716992.2
XP_006717055.1
11, 12
1546
XM_006716993.1
XP_006717056.1
11, 12
1546
XM_011518335.2
XP_011516637.1
11, 12
1546
XM_011518336.2
XP_011516638.1
11, 12
1546
XM_017014369.1
XP_016869858.1
11, 12
1546
XM_017014370.1
XP_016869859.1
11, 12
1546
XM_017014371.1
XP_016869860.1
11, 12
1546
XM_017014372.1
XP_016869861.1
11, 12
1546
XM_017014373.1
XP_016869862.1
11, 12
1546
GenBank mRNA
AB209124.1
-
10, 11
1231
AK096629.1
-
1, 2
2736
AK225179.1
-
11, 12
1455
AK295101.1
-
11, 12
1339
AK296326.1
-
11, 12
1513
AK308793.1
-
4, 5
347
AK316235.1
-
11, 12
1522
BC050279.1
-
11, 12
1522
5284480
BC063850.1
-
11, 12
1479
6184100
L07807.1
-
11, 12
1496
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor internalization
endocytosis
receptor-mediated endocytosis
endosome organization
sensory perception of sound
adult locomotory behavior
ephrin receptor signaling pathway
protein tetramerization
synaptic transmission, GABAergic
clathrin-mediated endocytosis
toxin transport
positive regulation of synaptic vesicle recycling
GTPase activity
protein binding
GTP binding
protein C-terminus binding
protein kinase binding
D2 dopamine receptor binding
protein complex binding
identical protein binding
poly(A) RNA binding
protein dimerization activity
nitric-oxide synthase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0