Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004423.3 | NP_004414.3 | 5 | 615 | ||
XM_005247172.2 | XP_005247229.1 | 5 | 800 | |||
XM_011512513.2 | XP_011510815.1 | 3 | 222 | |||
GenBank mRNA | AF006013.1 | - | 5 | 486 | ||
AK093469.1 | - | 5 | 607 | |||
AK304686.1 | - | 6 | 567 | |||
AL110278.1 | - | 2 | 76 | |||
BC018289.1 | - | 3 | 151 | |||
BC032459.1 | - | 5 | 543 | 5200610 | ||
D86963.1 | - | 5 | 615 | |||
U49262.1 | - | 5 | 596 | |||
U75651.1 | - | 5 | 868 |