Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ETS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256295.1 | NP_001243224.1 | 11 | 2996 | ||
NM_005239.5 | NP_005230.1 | 10 | 2826 | |||
XM_005260935.1 | XP_005260992.1 | 10 | 2899 | |||
XM_017028290.1 | XP_016883779.1 | 10 | 5496 | |||
GenBank mRNA | AA452482.1 | - | 1 | 273 |