Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EWSR1 SIRNA Assays ›
Gene Symbol:
EWSR1
Gene Name:
EWS RNA binding protein 1
Gene Aliases:
EWS, EWS-FLI1, bK984G1.4
Chromosome Location:
Chr.22: 29268009 - 29300525 on Build GRCh38
UniGene ID:
Hs.374477
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146641
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EWSR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163285.1
NP_001156757.1
4
527
NM_001163286.1
NP_001156758.1
4
527
NM_001163287.1
NP_001156759.1
4
527
NM_005243.3
NP_005234.1
4
527
NM_013986.3
NP_053733.2
4
527
XM_005261389.3
XP_005261446.1
4
282
XM_005261390.4
XP_005261447.1
4
282
XM_011529995.2
XP_011528297.1
4
257
XM_011529996.2
XP_011528298.1
4
257
XM_011529997.1
XP_011528299.1
4
257
XM_011529998.1
XP_011528300.1
4
257
XM_011529999.2
XP_011528301.1
4
257
XM_011530000.1
XP_011528302.1
4
257
XM_011530001.2
XP_011528303.1
4
257
XM_011530002.2
XP_011528304.1
4
257
XM_017028644.1
XP_016884133.1
4
282
XM_017028645.1
XP_016884134.1
4
282
XM_017028646.1
XP_016884135.1
4
257
XM_017028647.1
XP_016884136.1
4
282
XM_017028648.1
XP_016884137.1
4
282
XM_017028649.1
XP_016884138.1
4
257
XM_017028650.1
XP_016884139.1
4
257
XM_017028651.1
XP_016884140.1
4
282
XM_017028652.1
XP_016884141.1
4
257
XM_017028653.1
XP_016884142.1
4
282
XM_017028654.1
XP_016884143.1
4
257
XM_017028655.1
XP_016884144.1
4
282
XM_017028656.1
XP_016884145.1
4
282
XM_017028657.1
XP_016884146.1
4
243
XM_017028658.1
XP_016884147.1
4
257
XM_017028659.1
XP_016884148.1
4
282
XM_017028660.1
XP_016884149.1
4
282
XM_017028661.1
XP_016884150.1
4
243
XM_017028662.1
XP_016884151.1
4
282
XM_017028663.1
XP_016884152.1
4
282
XM_017028664.1
XP_016884153.1
4
237
XM_017028665.1
XP_016884154.1
4
237
XM_017028666.1
XP_016884155.1
4
243
GenBank mRNA
AK056309.1
-
4
220
AK056681.1
-
4
210
AK127624.1
-
236
AK225096.1
-
4
209
AK225102.1
-
4
221
BC000527.2
-
4
239
2961279
BC004817.1
-
4
218
3449145
BC011048.1
-
4
226
4340116
BC072442.1
-
3
205
6047805
CR456490.1
-
4
232
CX786322.1
-
4
257
DA013931.1
-
4
521
DA051447.1
-
4
225
DB051511.1
-
4
508
JX977847.1
-
4
527
X66899.1
-
4
242
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
nucleotide binding
RNA binding
protein binding
calmodulin binding
zinc ion binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0