Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EWSR1 SIRNA Assays ›
Gene Symbol:
EWSR1
Gene Name:
EWS RNA binding protein 1
Gene Aliases:
EWS, EWS-FLI1, bK984G1.4
Chromosome Location:
Chr.22: 29268009 - 29300525 on Build GRCh38
UniGene ID:
Hs.374477
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146642
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EWSR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163285.1
NP_001156757.1
5
676
NM_001163286.1
NP_001156758.1
5
676
NM_001163287.1
NP_001156759.1
5
676
NM_005243.3
NP_005234.1
5
676
NM_013986.3
NP_053733.2
6
694
XM_005261389.3
XP_005261446.1
5
431
XM_005261390.4
XP_005261447.1
5
431
XM_011529995.2
XP_011528297.1
5
406
XM_011529996.2
XP_011528298.1
5
406
XM_011529997.1
XP_011528299.1
5
406
XM_011529998.1
XP_011528300.1
5
406
XM_011529999.2
XP_011528301.1
5
406
XM_011530000.1
XP_011528302.1
5
406
XM_011530001.2
XP_011528303.1
5
406
XM_011530002.2
XP_011528304.1
5
406
XM_017028644.1
XP_016884133.1
5
431
XM_017028645.1
XP_016884134.1
5
431
XM_017028646.1
XP_016884135.1
5
406
XM_017028647.1
XP_016884136.1
5
431
XM_017028648.1
XP_016884137.1
5
431
XM_017028649.1
XP_016884138.1
5
406
XM_017028650.1
XP_016884139.1
5
406
XM_017028651.1
XP_016884140.1
5
431
XM_017028652.1
XP_016884141.1
5
406
XM_017028653.1
XP_016884142.1
5
431
XM_017028654.1
XP_016884143.1
5
406
XM_017028655.1
XP_016884144.1
5
431
XM_017028656.1
XP_016884145.1
5
431
XM_017028657.1
XP_016884146.1
5
4157
XM_017028658.1
XP_016884147.1
5
406
XM_017028659.1
XP_016884148.1
5
431
XM_017028660.1
XP_016884149.1
5
431
XM_017028661.1
XP_016884150.1
5
4157
XM_017028662.1
XP_016884151.1
5
431
XM_017028663.1
XP_016884152.1
5
431
XM_017028664.1
XP_016884153.1
5
386
XM_017028665.1
XP_016884154.1
5
386
XM_017028666.1
XP_016884155.1
5
4157
GenBank mRNA
AK056309.1
-
6
387
AK056681.1
-
5
359
AK225096.1
-
5
358
AK225102.1
-
5
370
BC000527.2
-
5
388
2961279
BC004817.1
-
5
367
3449145
BC011048.1
-
5
375
4340116
BC072442.1
-
4
354
6047805
BX648769.1
-
1
2524
CR456490.1
-
5
381
CX786322.1
-
5
406
DA051447.1
-
5
374
JX977847.1
-
5
676
X66899.1
-
5
391
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
nucleotide binding
RNA binding
protein binding
calmodulin binding
zinc ion binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-q6sk2:80/10.239.167.246:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline