Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: EIF2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201478.1 | NP_001188407.1 | 9 | 1083 | ||
NM_006893.2 | NP_008824.2 | 11 | 1455 | |||
XM_005277509.3 | XP_005277566.1 | 10 | 1220 | |||
XM_006711196.3 | XP_006711259.1 | 11 | 1326 | |||
XM_011509257.2 | XP_011507559.1 | 11 | 1370 | |||
XM_017000530.1 | XP_016856019.1 | 8 | 890 | |||
XM_017000531.1 | XP_016856020.1 | 8 | 848 | |||
XM_017000532.1 | XP_016856021.1 | 10 | 1176 | |||
GenBank mRNA | AB209364.1 | - | 2 | 3283 | ||
AF159586.1 | - | 6 | 554 | |||
AF220417.2 | - | 11 | 1363 | |||
AF262403.1 | - | 9 | 991 | |||
AK001852.1 | - | 11 | 1368 | |||
AK225259.1 | - | 11 | 1349 | |||
AL162001.1 | - | 6 | 661 | |||
BC001585.1 | - | 11 | 1353 | 3463026 | ||
BC039134.1 | - | 11 | 1306 | 4339127 | ||
BC058905.1 | - | 11 | 1320 | 6067337 |