Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DDR1 SIRNA Assays ›
Gene Symbol:
DDR1
Gene Name:
discoidin domain receptor tyrosine kinase 1
Gene Aliases:
CAK, CD167, DDR, EDDR1, HGK2, MCK10, NEP, NTRK4, PTK3, PTK3A, RTK6, TRKE
Chromosome Location:
Chr.6: 30880909 - 30900156 on Build GRCh38
UniGene ID:
Hs.631988
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146718
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DDR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001202521.1
NP_001189450.1
15
3058
NM_001202522.1
NP_001189451.1
15
2976
NM_001202523.1
NP_001189452.1
17
3555
NM_001297652.1
NP_001284581.1
17
3674
NM_001297653.1
NP_001284582.1
17
3476
NM_001297654.1
NP_001284583.1
18
3587
NM_001954.4
NP_001945.3
19
3594
NM_013993.2
NP_054699.2
19
3631
NM_013994.2
NP_054700.2
17
3432
XM_006715185.2
XP_006715248.1
18
3578
XM_011514882.2
XP_011513184.1
18
3596
XM_011514883.1
XP_011513185.1
20
3711
XM_011514884.1
XP_011513186.1
20
3787
XM_011514885.1
XP_011513187.1
18
3634
XM_011514886.1
XP_011513188.1
18
3499
XM_011514887.1
XP_011513189.1
18
3649
XM_011514888.2
XP_011513190.1
17
3485
XM_017011268.1
XP_016866757.1
18
3466
XM_017011269.1
XP_016866758.1
17
3538
GenBank mRNA
AB210021.1
-
18
3415
BC008716.2
-
19
3594
2964574
BC013400.2
-
19
3594
2964574
BC070070.1
-
17
3365
5262509
BM875592.1
-
1
240
L11315.1
-
19
3511
L20817.1
-
18
3551
L57508.1
-
17
3440
X74979.1
-
16
3306
Z29093.1
-
16
3599
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
regulation of cell-matrix adhesion
cell adhesion
embryo implantation
lactation
negative regulation of cell proliferation
regulation of extracellular matrix disassembly
smooth muscle cell migration
extracellular matrix organization
organ regeneration
collagen-activated tyrosine kinase receptor signaling pathway
peptidyl-tyrosine autophosphorylation
ear development
skin development
wound healing, spreading of cells
protein autophosphorylation
branching involved in mammary gland duct morphogenesis
mammary gland alveolus development
smooth muscle cell-matrix adhesion
transmembrane receptor protein tyrosine kinase activity
protein binding
collagen binding
ATP binding
protein tyrosine kinase collagen receptor activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline