Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CDH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016279.3 | NP_057363.3 | 7 | 1368 | ||
GenBank mRNA | AB035302.1 | - | 7 | 1353 | ||
AK307779.1 | - | 7 | 1369 | |||
BC107594.1 | - | 7 | 1330 | 40011942 | ||
BC113745.1 | - | 7 | 1330 | 40011941 |