Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEB SIRNA Assays ›
Gene Symbol:
NEB
Gene Name:
nebulin
Gene Aliases:
NEB177D, NEM2
Chromosome Location:
Chr.2: 151485334 - 151734487 on Build GRCh38
UniGene ID:
Hs.588655
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14683
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164507.1
NP_001157979.1
20
2027
NM_001164508.1
NP_001157980.1
20
2027
NM_001271208.1
NP_001258137.1
20
2027
NM_004543.4
NP_004534.2
20
2027
XM_005246590.2
XP_005246647.1
20
2014
XM_005246591.2
XP_005246648.1
20
2014
XM_005246592.2
XP_005246649.1
20
2014
XM_005246593.2
XP_005246650.1
20
2014
XM_005246594.2
XP_005246651.1
20
2014
XM_005246596.2
XP_005246653.1
20
2014
XM_005246597.2
XP_005246654.1
20
2014
XM_005246598.2
XP_005246655.1
20
2014
XM_005246599.2
XP_005246656.1
20
2014
XM_005246600.2
XP_005246657.1
20
2014
XM_005246601.2
XP_005246658.1
20
2014
XM_005246602.2
XP_005246659.1
20
2014
XM_005246603.2
XP_005246660.1
20
2014
XM_005246604.2
XP_005246661.1
20
2014
XM_005246606.2
XP_005246663.1
20
2014
XM_005246608.2
XP_005246665.1
20
2014
XM_005246610.2
XP_005246667.1
20
2014
XM_005246611.2
XP_005246668.1
20
2014
XM_005246612.2
XP_005246669.1
20
2014
XM_005246613.2
XP_005246670.1
20
2014
XM_005246615.2
XP_005246672.1
20
2014
XM_005246616.1
XP_005246673.1
20
2014
XM_005246617.2
XP_005246674.1
20
2014
XM_006712541.2
XP_006712604.1
20
2014
XM_006712542.2
XP_006712605.1
20
2014
XM_011511225.2
XP_011509527.1
20
2014
XM_011511226.2
XP_011509528.1
20
2014
XM_011511227.2
XP_011509529.1
20
2014
XM_017004177.1
XP_016859666.1
20
2014
XM_017004178.1
XP_016859667.1
20
2014
XM_017004179.1
XP_016859668.1
20
2014
XM_017004180.1
XP_016859669.1
20
2014
XM_017004181.1
XP_016859670.1
20
2014
XM_017004182.1
XP_016859671.1
20
2014
XM_017004183.1
XP_016859672.1
20
2014
XM_017004184.1
XP_016859673.1
20
2014
XM_017004185.1
XP_016859674.1
20
2014
GenBank mRNA
AK307607.1
-
20
2014
BC050436.1
-
8
723
X83957.1
-
7
2264
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle organ development
somatic muscle development
muscle filament sliding
regulation of actin filament length
actin binding
protein binding
structural constituent of muscle
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0