Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322019.1 | NP_001308948.1 | 2 | 188 | ||
NM_001322020.1 | NP_001308949.1 | 2 | 188 | |||
NM_004544.3 | NP_004535.1 | 2 | 188 | |||
NR_136155.1 | 2 | 188 | ||||
NR_136156.1 | 2 | 188 | ||||
NR_136158.1 | 2 | 188 | ||||
XM_011511228.2 | XP_011509530.1 | 2 | 175 | |||
GenBank mRNA | AB209440.1 | - | 2 | 109 | ||
AF087661.1 | - | 2 | 108 | |||
AF087854.1 | - | 2 | 127 | |||
AF453834.1 | - | 2 | 87 | |||
AK290778.1 | - | 2 | 110 | |||
BC003417.1 | - | 2 | 122 | 3451514 | ||
BC031332.1 | - | 2 | 93 | 4872196 | ||
BI551914.1 | - | 172 |