Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMK2G SIRNA Assays ›
Gene Symbol:
CAMK2G
Gene Name:
calcium/calmodulin dependent protein kinase II gamma
Gene Aliases:
CAMK, CAMK-II, CAMKG
Chromosome Location:
Chr.10: 73812501 - 73874591 on Build GRCh38
UniGene ID:
Hs.523045
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146843
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMK2G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204492.1
NP_001191421.1
21
2965
NM_001222.3
NP_001213.2
19
2833
NM_172169.2
NP_751909.1
21
2929
NM_172170.4
NP_751910.1
20
2902
NM_172171.2
NP_751911.1
21
3016
NM_172173.2
NP_751913.1
20
2860
XM_005270195.1
XP_005270252.1
22
3070
XM_005270197.1
XP_005270254.1
22
3034
XM_005270198.1
XP_005270255.1
22
3028
XM_005270199.1
XP_005270256.1
21
3001
XM_005270200.1
XP_005270257.1
21
2971
XM_005270201.1
XP_005270258.1
21
2965
XM_005270203.1
XP_005270260.1
21
2926
XM_005270205.1
XP_005270262.1
20
2887
XM_006717993.1
XP_006718056.1
23
3103
XM_006717996.1
XP_006718059.1
22
3040
XM_006717997.1
XP_006718060.1
22
2989
XM_006717998.1
XP_006718061.1
22
2947
XM_017016728.1
XP_016872217.1
21
2955
XM_017016730.1
XP_016872219.1
21
2884
XM_017016733.1
XP_016872222.1
20
2841
XM_017016735.1
XP_016872224.1
20
2857
XM_017016736.1
XP_016872225.1
21
2995
GenBank mRNA
AK026309.1
-
1
567
BC034044.1
-
21
2920
4813948
BC037928.1
-
1
405
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008770499, XM_008770497, XM_008770639, XM_008770495, XM_008770493, and others.
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
MAPK cascade
protein dephosphorylation
calcium ion transport
nervous system development
regulation of skeletal muscle adaptation
insulin secretion
cell differentiation
positive regulation of GTPase activity
protein autophosphorylation
protein oligomerization
regulation of calcium ion transport
interferon-gamma-mediated signaling pathway
regulation of cellular response to heat
regulation of relaxation of cardiac muscle
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
calcium-dependent protein serine/threonine phosphatase activity
Ras guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0