Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELF2 SIRNA Assays ›
Gene Symbol:
ELF2
Gene Name:
E74 like ETS transcription factor 2
Gene Aliases:
EU32, NERF, NERF-1A, NERF-1B, NERF-1a,b, NERF-2
Chromosome Location:
Chr.4: 139057717 - 139177218 on Build GRCh38
UniGene ID:
Hs.634040
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146875
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276457.1
NP_001263386.1
4
753
NM_001276458.1
NP_001263387.1
5
804
NM_001276459.1
NP_001263388.1
4
717
NM_006874.3
NP_006865.1
5
840
NM_201999.2
NP_973728.1
7
990
XM_005262803.2
XP_005262860.1
8
1299
XM_005262804.2
XP_005262861.1
8
1263
XM_005262805.2
XP_005262862.1
7
1176
XM_006714128.2
XP_006714191.1
7
1212
XM_006714129.3
XP_006714192.1
7
1078
XM_011531710.2
XP_011530012.1
8
1256
XM_011531711.2
XP_011530013.1
6
805
XM_011531712.2
XP_011530014.1
5
796
XM_011531714.2
XP_011530016.1
2
273
XM_017007856.1
XP_016863345.1
7
1098
XM_017007857.1
XP_016863346.1
7
1042
XM_017007858.1
XP_016863347.1
6
1047
XM_017007859.1
XP_016863348.1
7
1133
XM_017007860.1
XP_016863349.1
6
955
XM_017007861.1
XP_016863350.1
7
1083
GenBank mRNA
AF256222.1
-
7
942
AF256223.1
-
5
693
AK301307.1
-
5
685
AK303782.1
-
2
543
AK316455.1
-
5
715
BC034951.1
-
5
924
4821608
BC065025.1
-
4
682
6044351
BI770157.1
-
4
497
U43188.1
-
7
966
U43189.1
-
5
737
Back To Top
More Information
Panther Classification:
Molecular Function -
winged helix/forkhead transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
RNA polymerase II transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-9ljwx:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0