Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000018.3 | NP_000009.1 | 11 | 1308 | ||
NM_001033859.2 | NP_001029031.1 | 10 | 1242 | |||
NM_001270447.1 | NP_001257376.1 | 12 | 1244 | |||
NM_001270448.1 | NP_001257377.1 | 10 | 1383 | |||
XM_006721516.2 | XP_006721579.2 | 11 | 1261 | |||
XM_011523829.1 | XP_011522131.1 | 11 | 1261 | |||
XM_011523830.1 | XP_011522132.1 | 11 | 1261 | |||
GenBank mRNA | AK056083.1 | - | 10 | 1276 | ||
AK056956.1 | - | 6 | 654 | |||
AK058109.1 | - | 11 | 1190 | |||
AK097243.1 | - | 8 | 1990 | |||
AK293549.1 | - | 12 | 1244 | |||
BC000399.2 | - | 11 | 1246 | 2822593 | ||
BC012912.1 | - | 11 | 1230 | 4471040 | ||
BC020218.1 | - | 10 | 1150 | 5090104 | ||
D43682.1 | - | 11 | 1187 | |||
X86556.1 | - | 11 | 1239 |