Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATRX SIRNA Assays ›
Gene Symbol:
ATRX
Gene Name:
ATRX, chromatin remodeler
Gene Aliases:
ATR2, JMS, MRX52, MRXHF1, RAD54, RAD54L, SFM1, SHS, XH2, XNP, ZNF-HX
Chromosome Location:
Chr.X: 77504878 - 77786269 on Build GRCh38
UniGene ID:
Hs.533526
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146933
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATRX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000489.4
NP_000480.3
9
2668
NM_138270.3
NP_612114.2
8
2554
XM_005262153.4
XP_005262210.2
9
2665
XM_005262154.4
XP_005262211.2
10
2581
XM_005262155.4
XP_005262212.2
9
2769
XM_005262156.4
XP_005262213.2
8
2521
XM_005262157.4
XP_005262214.2
9
2464
XM_006724666.3
XP_006724729.1
8
2551
XM_006724667.3
XP_006724730.1
7
2407
XM_006724668.2
XP_006724731.1
9
2668
XM_017029601.1
XP_016885090.1
10
2578
XM_017029602.1
XP_016885091.1
9
2749
XM_017029603.1
XP_016885092.1
8
2518
XM_017029604.1
XP_016885093.1
9
2467
XM_017029605.1
XP_016885094.1
10
2682
XM_017029606.1
XP_016885095.1
8
2655
XM_017029607.1
XP_016885096.1
8
2652
XM_017029608.1
XP_016885097.1
7
2404
XM_017029609.1
XP_016885098.1
9
2568
XM_017029610.1
XP_016885099.1
9
2565
XM_017029611.1
XP_016885100.1
8
2320
GenBank mRNA
AB102641.1
-
9
2400
AB208928.1
-
9
2395
AB209545.1
-
1
293
AK308419.1
-
10
2520
U72936.2
-
10
2737
U72937.2
-
9
2615
U72938.2
-
8
2501
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
DNA methylation
DNA recombination
nucleosome assembly
DNA replication-independent nucleosome assembly
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
spermatogenesis
positive regulation of nuclear cell cycle DNA replication
covalent chromatin modification
DNA damage response, signal transduction by p53 class mediator
forebrain development
replication fork processing
positive regulation of telomere maintenance
DNA duplex unwinding
post-embryonic forelimb morphogenesis
multicellular organism growth
positive regulation of transcription from RNA polymerase II promoter
Sertoli cell development
protein localization to chromosome, telomeric region
seminiferous tubule development
cellular response to hydroxyurea
regulation of histone H3-K9 trimethylation
negative regulation of telomeric RNA transcription from RNA pol II promoter
positive regulation of telomeric RNA transcription from RNA pol II promoter
negative regulation of maintenance of mitotic sister chromatid cohesion, telomeric
DNA binding
DNA helicase activity
chromatin binding
helicase activity
protein binding
ATP binding
DNA translocase activity
methylated histone binding
histone binding
metal ion binding
chromo shadow domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0