Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000685.4 | NP_000676.1 | 3 | 1983 | ||
NM_004835.4 | NP_004826.5 | 3 | 1957 | |||
NM_009585.3 | NP_033611.1 | 2 | 1899 | |||
NM_031850.3 | NP_114038.4 | 4 | 2041 | |||
NM_032049.3 | NP_114438.2 | 3 | 1857 | |||
GenBank mRNA | BC022447.1 | - | 2 | 1867 | 4799755 | |
BC068494.1 | - | 3 | 1952 | 30337806 | ||
M87290.1 | - | 2 | 1875 | |||
S77410.1 | - | 3 | 1929 | |||
X65699.1 | - | 2 | 1833 |
Set Membership: |
![]() |