Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001087.4 | NP_001078.2 | 2 | 383 | ||
NM_001302545.1 | NP_001289474.1 | 2 | 386 | |||
GenBank mRNA | AB209790.1 | - | 2 | 262 | ||
AK131047.1 | - | 2 | 320 | |||
AK225078.1 | - | 2 | 291 | |||
BC020244.1 | - | 2 | 247 | |||
BC039866.1 | - | 2 | 311 | 6049156 | ||
BG534787.1 | - | 2 | 344 | |||
CN277005.1 | - | 2 | 343 | |||
CR456755.1 | - | 2 | 314 | |||
DA277259.1 | - | 2 | 383 | |||
DA465288.1 | - | 2 | 360 | |||
M95627.1 | - | 2 | 314 |