Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ANPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001150.2 | NP_001141.2 | 19 | 2843 | ||
XM_005254892.4 | XP_005254949.1 | 19 | 2851 | |||
XM_011521473.1 | XP_011519775.1 | 19 | 2994 | |||
GenBank mRNA | AB209918.1 | - | 19 | 2839 | ||
AK298134.1 | - | 13 | 1313 | |||
AK298242.1 | - | 13 | 1364 | |||
AK298339.1 | - | 14 | 1424 | |||
AK300946.1 | - | 15 | 1574 | |||
BC058928.1 | - | 18 | 2644 | 6020902 | ||
M22324.1 | - | 18 | 2671 | |||
X13276.1 | - | 18 | 2575 |