Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001647.3 | NP_001638.1 | 5 | 931 | ||
GenBank mRNA | BC007402.2 | - | 5 | 646 | 3050012 | |
BU195270.1 | - | 5 | 846 | |||
J02611.1 | - | 5 | 630 | |||
S80440.1 | - | 2 | 296 |