Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001656.3 | NP_001647.1 | 2, 3 | 334 | ||
NM_033227.2 | NP_150230.1 | 2, 3 | 334 | |||
NM_033228.2 | NP_150231.1 | 2, 3 | 334 | |||
XM_005248497.3 | XP_005248554.1 | 1, 2 | 167 | |||
XM_017009444.1 | XP_016864933.1 | 2, 3 | 318 | |||
GenBank mRNA | AF230397.1 | - | 2, 3 | 270 | ||
AF230398.1 | - | 2, 3 | 270 | |||
AF230399.1 | - | 2, 3 | 270 | |||
AK292814.1 | - | 2, 3 | 294 | |||
BC022510.1 | - | 2, 3 | 275 | 4811707 | ||
BI859501.1 | - | 2, 3 | 588 | |||
L04510.1 | - | 2, 3 | 270 |