Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001716.4 | NP_001707.1 | 2 | 1899 | ||
NM_032966.2 | NP_116743.1 | 1 | 1876 | |||
GenBank mRNA | BC110352.1 | - | 2 | 1813 | 5219901 | |
X68149.1 | - | 2 | 1805 | |||
X68829.1 | - | 1 | 1876 |