Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTNNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127384.2 | NP_001120856.1 | 9 | 1295 | ||
NM_013266.3 | NP_037398.2 | 9 | 1342 | |||
XM_017016151.1 | XP_016871640.1 | 10 | 1337 | |||
XM_017016152.1 | XP_016871641.1 | 12 | 1634 | |||
XM_017016153.1 | XP_016871642.1 | 10 | 1345 | |||
XM_017016154.1 | XP_016871643.1 | 5 | 713 | |||
XM_017016155.1 | XP_016871644.1 | 5 | 723 | |||
XM_017016156.1 | XP_016871645.1 | 5 | 600 | |||
XM_017016157.1 | XP_016871646.1 | 3 | 750 | |||
GenBank mRNA | AF091606.1 | - | 9 | 1320 | ||
AK289756.1 | - | 9 | 1231 | |||
AK313882.1 | - | 9 | 1345 | |||
BC065819.1 | - | 9 | 1238 | 6189390 |