Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MED24 SIRNA Assays ›
Gene Symbol:
MED24
Gene Name:
mediator complex subunit 24
Gene Aliases:
ARC100, CRSP100, CRSP4, DRIP100, MED5, THRAP4, TRAP100
Chromosome Location:
Chr.17: 40019097 - 40054636 on Build GRCh38
UniGene ID:
Hs.462983
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147535
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MED24
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079518.1
NP_001072986.1
12
1486
NM_001267797.1
NP_001254726.1
12
1464
NM_014815.3
NP_055630.2
13
1503
NR_052017.1
12
1429
XM_005257874.2
XP_005257931.1
13
1408
XM_006722204.2
XP_006722267.1
13
1408
XM_006722206.2
XP_006722269.1
13
1316
XM_006722207.2
XP_006722270.1
13
1284
XM_011525529.2
XP_011523831.1
14
1465
XM_011525530.2
XP_011523832.1
14
1341
XM_011525531.2
XP_011523833.1
13
1284
XM_011525532.2
XP_011523834.1
14
1373
XM_011525533.2
XP_011523835.1
14
1341
XM_011525534.2
XP_011523836.1
13
1284
XM_011525535.2
XP_011523837.1
13
1284
XM_011525536.2
XP_011523838.1
14
1341
XM_011525538.2
XP_011523840.1
13
1599
XM_017025458.1
XP_016880947.1
14
1373
XM_017025459.1
XP_016880948.1
13
1316
XM_017025460.1
XP_016880949.1
14
1373
XM_017025461.1
XP_016880950.1
14
1465
XM_017025462.1
XP_016880951.1
14
1373
XM_017025463.1
XP_016880952.1
14
1341
XM_017025464.1
XP_016880953.1
13
1284
XM_017025465.1
XP_016880954.1
13
1408
XM_017025466.1
XP_016880955.1
13
1316
XM_017025467.1
XP_016880956.1
13
1316
XM_017025468.1
XP_016880957.1
13
1284
XM_017025469.1
XP_016880958.1
13
1599
XM_017025470.1
XP_016880959.1
13
1599
XM_017025471.1
XP_016880960.1
13
1284
GenBank mRNA
AF055995.1
-
12
1190
AF277379.1
-
12
1190
AK022508.1
-
12
1255
AK291040.1
-
12
1235
AK293309.1
-
11
1025
AK300992.1
-
12
1108
AK303745.1
-
12
1199
BC011375.1
-
12
1465
3866038
D50920.1
-
13
1263
EU392550.1
-
6
368
Back To Top
More Information
Panther Classification:
Molecular Function -
general transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
protein ubiquitination
histone acetylation
stem cell population maintenance
intracellular steroid hormone receptor signaling pathway
androgen receptor signaling pathway
positive regulation of transcription, DNA-templated
protein heterooligomerization
RNA polymerase II transcription cofactor activity
transcription cofactor activity
histone acetyltransferase activity
receptor activity
protein binding
ligand-dependent nuclear receptor transcription coactivator activity
vitamin D receptor binding
thyroid hormone receptor binding
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0